Drukuj
Akcesoria do transiluminatorów
Dostawca: ELMA SCHMIDBAUER
1001476 1001357 1001361 1001461 1004536
142-2227EA 0 PLN
142-2227 142-2223 142-2224 142-2226 ELMA1004536
Akcesoria do transiluminatorów
Łaźnie ultradźwiękowe
The X-Tra series baths have two levels of cleaning. Powerclean for intensive cleaning and Softclean for careful cleaning of sensitive materials. By using stainless steel cavitation, the baths have a very long lifetime, which is further supported by a three year warranty.
  • Sweep function, for a homogeneous ultrasound distribution
  • Additional high performance oscillation systems
  • High heater for short heating times, dry-running protection
  • Fast drainage