Drukuj
Akcesoria do Turbiquant®
Dostawca: Merck
1.18336.0001 1.18320.0001 1.18344.0001
1.18336.0001EA 1400 PLN
1.18336.0001 1.18320.0001 1.18344.0001
Akcesoria do Turbiquant®
Mętnościomierze
In addition to spare cuvettes and light sources, the pour and flow cells have been developed to improve efficiency. The pour through assembly provides a single one-time cuvette alignment and simplified rinsing.
Continuous flow through measurement is possible with the Turbiquant 3000 T/3000 IR by using the flow assembly together with the periodically measured output via printer or PC.