Drukuj
Butelki, kwadratowe, wąska szyjka, prostokątne, HDPE, z gwintem, seria 310
Dostawca: Retsch GmbH
32.354.0001 32.354.0004 32.902.0001 32.050.0001 32.354.0002 32.365.0001 32.902.0002
510-1983EA 247 PLN
510-1983 411-3317 510-1982 510-1785 510-1786 510-1787 510-1788
Butelki, kwadratowe, wąska szyjka, prostokątne, HDPE, z gwintem, seria 310
Sita Sita testowe
These accessories are designed for products that are difficult to separate, which are not suitable for wet sieving.