Drukuj
Sita laboratoryjne, 200×50 mm
Dostawca: Retsch GmbH
60.131.000020 60.131.000025 60.131.000032 60.131.000036 60.131.000038 60.131.000040 60.131.000045 60.131.000050 60.131.000053 60.131.000056 60.131.000063 60.131.000071 60.131.000075 60.131.000080 60.131.000090 60.131.000100 60.131.000106 60.131.000112 60.131.000125 60.131.000140 60.131.000150 60.131.000160 60.131.000180 60.131.000200 60.131.000212 60.131.000224 60.131.000250 60.131.000280 60.131.000300 60.131.000315 60.131.000355 60.131.000400 60.131.000425 60.131.000450 60.131.000500 60.131.000560 60.131.000600 60.131.000630 60.131.000710 60.131.000800 60.131.000850 60.131.000900 60.131.001000 60.131.001120 60.131.001180 60.131.001250 60.131.001400 60.131.001600 60.131.001700 60.131.001800 60.131.002000 60.131.002240 60.131.002360 60.131.002500 60.131.002800 60.131.003150 60.131.003350 60.131.003550 60.131.004000 60.131.004500 60.131.004750 60.131.005000 60.131.005600 60.131.006300 60.131.006700 60.131.007100 60.131.008000 60.131.009000 60.131.009500 60.131.010000 60.131.011200 60.131.012500 60.131.014000 60.131.016000 60.131.018000 60.131.019000 60.131.020000 60.131.022400 60.131.025000 60.131.026500 60.131.028000 60.131.031500 60.131.035500 60.131.037500 60.131.040000 60.131.045000 60.131.050000 60.131.053000 60.131.056000 60.131.063000 60.131.071000 60.131.075000 60.131.080000 60.131.090000 60.131.100000 60.131.106000 60.131.112000 60.131.125000 60.131.013200
510-4501EA 4820 PLN
510-4501 510-4502 510-4503 510-4504 510-4505 510-4506 510-4507 510-4508 510-4509 510-4510 510-4511 510-4512 510-4513 510-4514 510-4515 510-4516 510-4517 510-4518 510-4519 510-4520 510-4521 510-4522 510-4523 510-4524 510-4525 510-4526 510-4527 510-4528 510-4529 510-4530 510-4531 510-4532 510-4533 510-4534 510-4535 510-4536 510-4537 510-4538 510-4539 510-4540 510-4541 510-4542 510-4543 510-4544 510-4545 510-4546 510-4547 510-4548 510-4549 510-4550 510-4551 510-4552 510-4553 510-4554 510-4555 510-4556 510-4557 510-4558 510-4559 510-4560 510-4561 510-4562 510-4563 510-4564 510-4565 510-4566 510-4567 510-4568 510-4569 510-4570 510-4571 510-4572 510-4574 510-4575 510-4576 510-4577 510-4578 510-4579 510-4580 510-4581 510-4582 510-4583 510-4584 510-4585 510-4586 510-4587 510-4588 510-4589 510-4590 510-4591 510-4592 510-4593 510-4594 510-4595 510-4596 510-4597 510-4598 510-4599 510-4573
Sita laboratoryjne, 200×50 mm
Sita Sita testowe
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. Available in three of the most commonly used sizes. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Special certificates available on request. Contact your local Avantor sales office for further details.
Other mesh sizes available on request.
Other mesh sizes available on request.
Certyfikaty: ISO 3310-1:2001-09