Drukuj
Wytrząsarki Vortex, Vortex-Genie®
Dostawca: SCIENTIFIC INDUSTRIES ELMIS
SI-0256 SI-0156 SI-T256 SI-0276 SI-0176 SI-T276 SI-0266 SI-0166
444-5900EA 2970 PLN
444-5900 444-9173 444-9174 444-9182 444-9183 444-9184 444-0486 444-0485
Wytrząsarki Vortex, Vortex-Genie®
Wytrząsarki Wytrząsarki typu vortex
Rugged and reliable vortex mixers with solid metal casing and rubber feet that prevent them 'walking'. Vortex-Genie® 1 features touch-on operation and fixed speed, it is ideal for aggressive vortexing of difficult samples.
  • Wide range of attachments available
  • Vortex-Genie® 2T timer function; touch-on 1 to 60 seconds, hands-free 1 to 60 minutes or continuous
  • Can be used in cold room or incubator
Vortex-Genie® 2 has variable speed for precise mixing from gentle to vigorous, it offers hands-free or touch-on control. Mixing action prevents spilling, even if tubes are uncapped.
Vortex-Genie® 2T has the same features as Vortex-Genie® 2, and in addition includes an integral timer function, it can also be used on continuous untimed mode.
Dodatkowe informacje: Vortex-Genie® 1 is supplied with quick change standard pop-off cup for single test tubes. Optional 76 mm platform suitable for beakers, flasks and multiple tests tubes is available. Vortex-Genie® 2 and 2T are supplied with standard pop-off cup for single test tubes and 76 mm platform suitable for beakers, flasks and multiple tests tubes.
Uwaga: Depending on speed
Multiple sample starter set consists of a 152 mm platform, a foam insert for 60 microtubes and a foam insert for 1 standard microplate
Large sample set is ideal for mixing beakers, flasks and large tubes. The set consists of a recessed platform, 2 elastic bands, a blank foam insert, a foam insert for 4×29 to 37 mm tubes, a foam insert for 8×14 to 29 mm tubes and a foam insert for 12×9 to 16 mm tubes
Small ampoule/tube attachment enables vigorous end-to-end mixing for up to 4×15 to 17 mm Ø tubes or ampoules
Large ampoule/tube attachment enables vigorous end-to-end mixing for up to 4×10 to 17 mm Ø tubes or ampoules