Drukuj
Pipety jednokanałowe automatyczne, Transferpettor™
Dostawca: Brand
701807 701812 701817 701822 701842 701844 701853 701858 701863 701864 701868 701873 701878 702804 702806 702810 702812
613-3354EA 2110 PLN
613-3354 613-3355 613-3356 613-3357 613-3358 613-3359 613-3362 613-3363 613-3364 613-3365 613-3366 613-3367 613-3368 613-3379 613-3380 613-3381 613-3382
Pipety jednokanałowe automatyczne, Transferpettor™
Pipety Pipety i dozowniki seryjne
Ideal for pipetting highly viscous media up to 140000 mm²/s, or with density up to 13,6 g/cm³ (protein solutions, oils, fat, mercury, sulphuric acid), high vapour pressure media up to 500 mbar (alcohols, ethers, hydrocarbons), or media with a tendency to foam (tensioactive solutions).
  • For pipetting media that present problems for air displacement pipettes
  • Positive displacement ensures that results are reproducible regardless of pipetting speed and ambient conditions
  • The movable piston is in direct contact with the liquid and removes the last drop from the walls of the capillaries or tips
  • Residual moisture in the capillaries or tips is minimal and may be disregarded except in the case of infectious or radioactive media
  • DE-M marking, with performance certificate
Informacje do zamówienia: Must be used with capillaries, tips and pistons for Transferpettor™.