Drukuj
Wytwornica lodu płatkowego, seriaAF
Dostawca: SCOTSMAN
AF103 AF156 AF124
474-0110EA 21300 PLN
474-0110 474-0112 474-0111
Wytwornica lodu płatkowego, seriaAF
Wytwornice lodu
The AF self contained ice flakers are air-cooled and manufactured from 304 stainless steel.
  • Integral ice flake storage bin
  • Constructed of high grade stainless steel
  • Ideal for medical and laboratory use
All the models are automatic and produce small irregular flakes of ice with an even melting rate. The flakes are easily moulded to fit into any shape. The ice flakers require a 19 mm 'washing machine' type tapped mains water supply, a correctly rated power supply and a 38 mm vented drain (ideally no higher than the base of the machine), within one metre of the machine's intended position.
* Bin capacities are based on volumetric sizes of storage bins, actual capacity may be less due to ice 'pyramiding' as it falls into the bin.
** Add 120 mm for the legs, if required.
Informacje do zamówienia: Please contact Avantor prior to purchase if connection to a de-ionised water supply is preferred. Please note that water filtration systems are available for these ice makers. They are designed to reduce unscheduled service calls associated with the equipment's internal water system, this includes lime scale build-up, corrosion, sediment, bacteria etc. Please contact Avantor for full details.
Dodatkowe informacje: All devices are delivered without a plug.