Drukuj
Analogowy termohigrometr
Dostawca: Thies Clima
1.0169.42.058 1.0165.42.058
628-1139EA 1450 PLN
628-1139 628-2226
Analogowy termohigrometr
Higrometry Higrometry analogowe
These combined indicating instruments measure ambient temperature and relative humidity simultaneously.
  • Round design
  • Black figures on white background
  • Two coloured arrows for temperature and humidity
Certyfikaty: Compliant with DIN 50014.