Drukuj
Homogenizatory laboratoryjne, Stomacher® 80
Dostawca: Seward (FermionX)
0080/000/EU 0080/000/EU/M
432-3110EA 16550 PLN
432-3110 432-3155
Homogenizatory laboratoryjne, Stomacher® 80
Homogenizatory Homogenizatory laboratoryjne
Stomacher® lab blenders offer fast reliable blending and homogenisation of a broad range of laboratory samples. The Stomacher® machines work by the action of twin-reciprocating paddles on the sample contained inside a sterile plastic bag.
  • 3 year warranty, 10 year service and spare parts
  • Versatile and efficient blending and homogenisation of laboratory samples
  • Safe containment of sample in sterile plastic bag prevents cross-contamination
  • Machines do not need to be cleaned and sterilised between samples
  • No temperature increase
Zobacz ceny
SPECYFIKACJE
Typ Stomacher® 80
Prędkość 230 / 265 / 300 ±5 min⁻¹
Environmentally Preferable
Czasomierz 30 / 60 / 120 / continuous
Wymiary - szer. x gł. x wys. 290×320×180 mm
Waga 15 kg