Drukuj
Stół do mikro wag i wag analitycznych
Dostawca: Sartorius Balances
YWT03 YWT09
611-1131EA 14430 PLN
611-1131 611-1702
Stół do mikro wag i wag analitycznych
Meble Stoły
High performance micro and analytical balances should be placed in an environment where interfering vibration is at a minimum, to give fast stabilisation of the reading. The YWT09 weighing table features a sturdy frame with a granite stone slab insert.
  • Easy to clean surfaces
  • Fast stabilisation times
Informacje do zamówienia: Assembly is not included.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable