Drukuj
Aparat do destylacji behrotest® Kompact
Dostawca: BEHR
B00217712 B00217714 B00217716 B00217718 B00217720 B00217736 B00217706 B00217738 B00217741
705-0003EA 4780 PLN
705-0003 705-0004 705-0005 705-0006 705-0007 BEHRB00217736 705-0000 BEHRB00217738 BEHRB00217741
Aparat do destylacji behrotest® Kompact
Systemy do mineralizacji Stacje mineralizacji
Complete sets of equipment for preparation and distillation for commonly required analyses of solid and aqueous samples.
  • Simple and convenient to use: All apparatus is delivered already mounted on stands with tubing connections pre-installed - no more tedious fiddling with clamps and mountings
  • Space saving: All apparatus is mounted on a compact 23×27 cm base plate, including the units for dual sampling
  • Analytical reliability: All components comply with the applicable official analytical procedures and safety standards