Drukuj
Rejestratory temperatury, SC3
Dostawca: DICKSON
SC377 SC386 SC387 SC367
620-0256EA 3380 PLN
620-0256 620-0257 620-0258 620-0255
Rejestratory temperatury, SC3
Przyrządy rejestrujące Nagrywarki / rejestratory
These recorders are used to record temperature simply and reliably. They provide the users with immediate access to data and suit applications in manufacture of pharmaceutical, medical and food products and vaccine storage.
  • Withstand the rigours of transport and storage
  • Accuracy and resolution conform to quality controls
  • For recordings over 24 hours or seven days
W×D×H: 93×93×57 mm
Weight: 227 mm
Dodatkowe informacje: Supplied with a single AA battery with a service life of two years and a red pen. Other diagrams are available on request.