Drukuj
Wagi przemysłowe, Valor™ 3000 Xtreme
Dostawca: OHAUS
71210135 71210133 71210134
611-5322EA 1555 PLN
611-5322 611-5320 611-5321
Wagi przemysłowe, Valor™ 3000 Xtreme
Wagi
A compact, portable scale suitable for use in a wide range of applications including the food industry where dry or wet conditions are normal. The Valor™ Xtreme features a stainless steel housing, a removable stainless steel weighing pan and a bright backlit display. The software applications are designed for simple use and maximum throughput and include percent weighing and specific gravity. User adjustable filtering and stability settings allow the Valor™ 3000 Xtreme to operate accurately in draughty and unstable conditions. Digital external span calibration with specified weights. OIML EC type approved models, denoted by M suffix e.g. VX31XW6M, are also available on request.
  • Portable operation in battery power mode
  • IP 65 water resistant - for simple wash down in battery mode
  • SmarText™ interface for user friendly simple text prompts
  • QuadraStance™ adjustable feet and level bubble for stability
  • Overload (x10 capacity) and shock protection for tough environments
Dodatkowe informacje: Supplied with AC adapter as standard. Batteries (4× C-type/ LR14) are not included.