Drukuj
Anemometry wiatraczkowe, 410-1/2
Dostawca: TESTO
0560.4101 0560.4102
620-3307EA 562 PLN
620-3307 620-3308
Anemometry wiatraczkowe, 410-1/2
Wiatromierze
Handy, small and simple, these air velocity meters can measure temperature and wind speed, as well as humidity (620-3308 only). They can also perform wind-chill measurements.
  • Enclosed measurement with 40 mm vane
  • Minimum/maximum values
  • Automatic off function
  • Data 'HOLD' recall
  • Backlit display
W×D×H: 133×46×25 mm
Weight: 110 g
Dodatkowe informacje: Supplied with two alkaline batteries, protective cap, wrist strap and belt holder.