Drukuj
Chusteczki do ogólnych zastosowań, WYPALL* X70
Dostawca: KIMBERLY CLARK
8381 7867 8387 8389
115-1281EA 923 PLN
115-1281 115-1278 115-1282 KIMB8389
Chusteczki do ogólnych zastosowań, WYPALL* X70
Chusteczki Ściereczki wielorazowe Ściereczki uniwersalne
Made from fast-absorbing HYDROKNIT* material, a hydroentangled cellulose/polypropylene blend without adhesives or chemical binders. For use in heavy manufacturing, metal fabrication, and processing plants for chemicals, refining, rubber, and plastics.
  • Durable, rag-tough construction and a textured surface, yet soft and pliable
  • Rags are reusable and will not fall apart when wet
  • Long lasting, on-the-job performance allows extended usage, reducing cost and waste
  • Large roll (115-2100) can be used with dispensers 115-0045, 115-1533 and 115-2038
Colour: White.
Colour: White
Uwaga: *The product(s) on this page are outside the scope of the EU Directive 2019/904.