Drukuj
Chusteczki, Tork
Dostawca: Essity
101250 131135 121206 150109 120155 100134 66310 130034 130100 128207 130041 130045 130050 130052 130070 130081 130062 128208 129239 129237 130060 129262 130080 101221 120123 100130 130042 130044 473382
115-2409EA 697 PLN
115-2409 115-0219 115-0213 115-1610 115-0210 115-0209 115-1697 115-0333 115-0334 115-1598 115-3109 115-0637 115-0215 115-0636 115-3112 115-0633 115-0635 115-1599 115-1602 115-1601 115-1607 115-1603 115-0656 115-1263 115-1264 115-1394 115-1604 115-1606 SCAA473382
Chusteczki, Tork
Chusteczki Chusteczki i ręczniki jednorazowe Chusteczki do ogólnych zastosowań
Centrefeed rolls and wipes for Tork dispenser systems available in various qualities.
  • Tork Heavy-Duty: Soft premium quality for heaviest wiping tasks - with a better, faster absorption than other towels and wipes
  • Tork Plus: Improved performance tissues and wipes that meet or exceed basic requirements - with a good absorption, brightness and strength
  • Tork Basic: Standard quality tissues and wipes for basic demands