Drukuj
Papier Josepha
Dostawca: PRAT DUMAS
D000407 D004407 D500407
291-0014EA 1410 PLN
291-0014 291-0015 PRATD500407
Papier Josepha
Chusteczki Chusteczki i ręczniki jednorazowe Chusteczki do delikatnych powierzchni
Joseph paper is ideal for precise cleaning applications. Superior quality for wiping or packaging delicate objects.
  • Low linting
  • Absorbs liquids easily and quickly
L×W (mm): 350×500
Weight: 30 g/m²