Drukuj
Chusteczki do ogólnych zastosowań, KIMTECH PURE*
Dostawca: KIMBERLY CLARK
7624 7623
115-2034EA 1230 PLN
115-2034 115-2033
Chusteczki do ogólnych zastosowań, KIMTECH PURE*
Chusteczki Ściereczki wielorazowe Ściereczki uniwersalne
KIMTECH PURE* cleaning wipes are manufactured using 100% virgin polypropylene. They are strong and absorbent allowing fast and efficient wiping and can be re-used.
  • Highly absorbent and give a softer feel
  • Chemically inert - they won't break down when used with solvents
  • Low linting, making them suitable for critical wiping tasks
  • Woven embossed pattern, which picks up and retains dirt making cleaning quicker and easier
Roll can be used with dispenser 115-0045, 115-1533 or 115-2038.
Colour: White