Drukuj
Pierścienie obciążające do naczyń, SP Bel-Art
Dostawca: Bel-Art Products, a Part of SP
F183080500 F183081000 F183082000 F183084000 183086500
214-0004EA 401 PLN
214-0004 214-0005 214-0006 214-0007 BELA183086500
Pierścienie obciążające do naczyń, SP Bel-Art
Odważniki Pierścienie obciążające
Lead core, with green vinyl coating. Open design (C shaped) is suitable for vessels with lateral connections. For stabilising glass or plastic vessels on shakers or in water and oil baths, etc. The weights prevent tipping. The coating prevents the vessels from being scratched or causing damage to other surfaces.
  • Good chemical resistance
  • High temperature resistance
  • Available in various sizes for use with a range of vessel sizes
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable