Drukuj
Zamknięcia do membran dializacyjnych, Spectra/Por®
Dostawca: Spectrum Laboratories
132734 132735 132736 132737 132738 132742 132743 132744 132745 132746 132748 132749 132750 132751 132752 132753 132760 132762 132764 132766 142737 142738 142836 142837 142838 142936 142937 142938 142736 132739 132747 142739 142839 142939
734-0689EA 903 PLN
734-0689 516-0189 734-0690 516-0190 734-0691 734-0692 734-0693 734-0694 734-0695 734-0696 516-2000 516-2001 516-0191 516-0192 516-2002 516-0193 514-0621 734-0699 734-0700 734-0701 514-0623 734-0738 734-0740 514-0625 734-0741 516-0196 516-0197 734-0743 514-0622 SPEC132739 SPEC132747 SPEC142739 SPEC142839 SPEC142939
Zamknięcia do membran dializacyjnych, Spectra/Por®
Zaciski Zaciski do węży
Standard closures
PP, autoclavable
Only for regenerated cellulose membranes
Lighter than water, normally used in combination with weighted closures
Choose a closure 4 to 10 mm larger than the flat width of the tubing
Weighted closures
PP with fluorocarbon coated stainless steel bar
Only for regenerated cellulose membranes
Used in combination with a standard closure for a vertical use of the dialysis tubing
Choose a closure 4 to 10 mm larger than the flat width of the tubing
Magnetic weighted closures
PP with PTFE coated magnetic weight
Only for regenerated cellulose membranes
Used in combination with a standard closure for a vertical use of the dialysis tubing and a magnetic stirrer for continuous stirring of the tubing
Choose a closure 4 to 10 mm larger than the flat width of the tubing
Choose a closure 4 to 10 mm larger than the flat width of the tubing.