Drukuj
Probówki ze szlifem i korkiem
Dostawca: Brand
114521 114508 114516 114524
212-1646EA 111 PLN
212-1646 212-1642 212-1644 212-1648
Probówki ze szlifem i korkiem
Probówki Probówki
DURAN®, borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Standard ground joint and PP stopper. Easy to read white graduations and labelling.