Drukuj
Akcesoria do dygestorium, seria Cerlaton
Dostawca: LUDI
184110621 184110622 184110623 184110731 184110732 184110733
391-0094EA 127 PLN
391-0094 LUDI184110622 LUDI184110623 391-5930 391-5932 391-5931
Akcesoria do dygestorium, seria Cerlaton
Nakrętki Nakrętki do probówek
Aluminium.
  • Made of anodised aluminium with inner stainless steel spring
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
With grip. Cap for sterile but not hermetic closing of test tubes and other vessels.