Drukuj
Probówki mikrocentrifugi z przymocowanymi zakrętkami, 0,5/1,5/2,0 ml
Dostawca: Brand
780710 780711 780712 780713 780714
211-3211EA 1220 PLN
211-3211 211-3212 211-3213 211-3214 211-3215
Probówki mikrocentrifugi z przymocowanymi zakrętkami, 0,5/1,5/2,0 ml
Probówki Probówki do mikrowirówek
PP, graduated, autoclavable, PP screw cap with silicone seal.
  • Can withstand centrifugation to 17000×g
  • Ideal for storage of serum and blood samples as well as for boiling and centrifugation
  • Positive sealing screw cap with silicone seal
  • Fast opening and closing with 1¹/₄ turn of cap
  • Operating range: −196 to +121 °C, suitable for freezing samples in the gas phase of liquid nitrogen at −196 °C
*without graduations
Informacje do zamówienia: Coloured cap inserts for identification are available separately.