Drukuj
Wózki transportowe
Dostawca: Metro International
BC2636-2D-G BC2636-3D-G BC2030-2DG BC2030-2DBL BC2636-3DBL
139-9993EA 2350 PLN
139-9993 139-9994 139-0054 METOBC2030-2DBL METOBC2636-3DBL
Wózki transportowe
Wózki
Chrome plated steel frame with HDPE trough shaped trays.
  • Stable, tough and versatile, easy to clean
  • Load capacity: Max. 65 kg for each tray; combined 180 kg
  • Rubber wheels to protect floors
The troughs are 70 mm deep, enabling liquids to be transported safely retaining any spillage. The height of the middle tray can be adjusted. Available in two sizes, with two or three trays.