Drukuj
Tace laboratoryjne
Dostawca: BOCHEM
8702 8703 8705 8706 8700 8704 8701
216-1362EA 747 PLN
216-1362 216-0048 216-0049 216-1364 216-1361 216-1363 216-0047
Tace laboratoryjne
Tace/Naczynia/Parownice Tace/naczynia laboratoryjne
Stainless steel, ¹⁸/₁₀.
  • Rounded corners for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, excellent thermal resistance
Multipurpose trays for a wide range of uses. Ideal as developing trays.