Drukuj
Termometry ogólnego przeznaczenia w powłoce PTFE
Dostawca: Alla France
512A110IMTJ-B 512250IMPJ-B 512250IMPJ-B-T 5122P110-T-QP 5182P110-T-QP 512A110IMTJ-R
620-0968EA 133 PLN
620-0968 620-0291 620-0292 620-0284 620-0294 620-1809
Termometry ogólnego przeznaczenia w powłoce PTFE
Termometry Analogue Thermometers
These thermometers are sheathed to prevent any contamination by pieces of glass or the liquid if they break. The range includes models with chemically resistant PTFE sheath.
  • Yellow background
  • Maximum safety
  • Biotemp® range with PTFE protective sheath
Ø: 6,0 to 7,0 mm
Organic liquid, toluen or red liquid