Drukuj
Ochraniacze powierzchni
Dostawca: PRAT DUMAS
N042205 N042205GRIS N042205PLIE25 N042205PLIE
115-2319EA 1130 PLN
115-2319 115-0654 115-0655 PRATN042205PLIE
Ochraniacze powierzchni
Ochraniacze powierzchni Ochraniacze powierzchni
These protection papers are available in different formats. They can be used for bench protection as well as for the packaging of glass parts.
  • Sheets are available flat or folded
  • Suitable for food contact
  • Cellulose and cotton
Weight: 65 g/m²
Porosity: 7 to 10 µm
Weight: 65 g/m²
Thickness: 0,16 mm
Porosity: 7 - 10 µ