Drukuj
Płytki HPTLC z cząstkami sferycznymi, LiChrospher®
Dostawca: Merck
1.05586.0001 1.05646.0001 1.15445.0001
1.05586.0001EA 1830 PLN
1.05586.0001 1.05646.0001 1.15445.0001
Płytki HPTLC z cząstkami sferycznymi, LiChrospher®
Płytki do TLC
HPTLC LiChrospher® plates are based on spherical-shaped silica 60 with a particle size of 7 µm, and narrow particle size distribution similar to the phases used in HPLC. LiChrospher® HPTLC plates possess a selectivity that is comparable to respective HPTLC plates, however, plate height and separation numbers are further improved resulting in shorter analysis times.
  • 20% reduced running times
  • Highly compact spots or zones
  • Lower detection limits and better resolution
LiChrospher® HPTLC plates are especially suitable for separations of highly complex, low concentration samples, such as:
Trace analysis of pesticide mixtures
Assay of pharmaceutical compounds