Drukuj
Płytki do PCR, 96-dołkowe, SuperPlate™
Dostawca: Thermo Fisher Scientific
AB-2400 AB-2400/W AB-2800 AB-2800/W
732-1060EA 914 PLN
732-1060 732-1063 732-1049 732-1052
Płytki do PCR, 96-dołkowe, SuperPlate™
Mikropłytki Płytki do PCR
PP. Thermo Scientific SuperPlate™ provides four times more rigidity for superior robotic handling. Three versions are available, all with ultra-rigid SBS footprint for compatibility with automated systems, and thin walled round bottom wells for optimal heat transfer and maximum sample recovery.
  • Enhanced thermal stability to minimise warping during thermal cycling and heat sealing
  • Black lettering for quick referencing of wells
  • Maximum well volume 0,2 ml when used with adhesive heat seals
  • Skirted version is stackable and compatible with automated systems
  • Available in white for qPCR applications