Drukuj
Cylindry pomiarowe, wysokie, klasa A, szkło borokrzemianowe, BLAUBRAND® ETERNA
Dostawca: Brand
32762 32764 32705 32708 32720 32728 32738 32748 32754 2.32762
612-5037EA 420 PLN
612-5037 612-9268 612-5030 612-5031 612-5032 612-5033 612-5034 612-5035 612-5036 612-4898
Cylindry pomiarowe, wysokie, klasa A, szkło borokrzemianowe, BLAUBRAND® ETERNA
Cylindry Cylindry miarowe
Borosilicate 3.3 glass, tall form, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, with spout
  • Ring marks at major graduations, marks and inscriptions in ETERNA amber stain
DIN EN ISO 4788
Certyfikaty: DIN EN ISO 4788
Informacje do zamówienia: Supplied with a batch certificate. Measuring cylinders are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.