Drukuj
Półmaski 620 N i 620 S
Dostawca: BartelsRieger Atemschutztechnik
111-3067EA 444 PLN
111-3067 111-3050
Półmaski 620 N i 620 S
Maski oddechowe Maski wielokrotnego użytku Półmaski
This reusable half-mask ensures safe breathing when sanding and spraying in industry, as well as during pest control and many other tasks. As well as being used as pure filter units, the masks can also be used as ventilation connections for light airline respirators due to their EN 148-1-compliant standard round-thread connections.
  • The mask body features an optimised design suitable for any face shape
  • With roller straps for quick adjustment and retensioning of the mask
  • Two exhalation valves guarantee low exhalation resistance
  • Choice of skin-friendly, highly durable rubber or silicone, specifically for allergy sufferers
Certyfikaty: EN 140, 148