Drukuj
Półmaski filtrujące cząstki, jednorazowe, FFP1/FFP2/FFP3, serii 5000
Dostawca: HONEYWELL SAFETY
1005580 1005113 1005582 1005586-V2 1005099 1005602-V2 1005588 1005593 1005591 1005595 1005598 1005600 1005122
111-3546EA 189 PLN
111-3546 111-3547 111-3552 111-3562 111-3563 111-3571 111-1275 111-3570 111-3569 111-3572 111-3574 111-3576 DALL1005122
Półmaski filtrujące cząstki, jednorazowe, FFP1/FFP2/FFP3, serii 5000
Maski oddechowe Maski jednorazowe Maski filtrujące cząstki
  • Ultra-soft seal providing a soft textile sensation on the skin, absorbing perspiration, ensuring a good seal; hypoallergenic qualities
  • 'Butterfly' exhalation valve offering maximum frontal protection on the valve membrane while also providing minimal exhalation resistance
  • Latex-free, PVC-free and silicone-free materials
  • Dolomite-tested: Test demonstrating the respirator's resistance to clogging over time; ensures better breathing comfort and less worker fatigue
Colour codes: FFP1D - yellow, FFP2D - green, FFP3D - orange, FFP1D-OV - brown for organic vapours (for FFP1D, FFP2D), FFP1D-AV - blue for acid gases and vapours (for FFP1D, FFP2D).
EN 149:2001
Certyfikaty: EN 149:2001
Uwaga: "NR" marking = non reusable.