Drukuj
Ultra-Micro Cells do pomiarów fluorescencji
Dostawca: WTW
106667 106663 106653 106651 106675 106665 106666 106671 106661 106655 106649
662-7379EA 7180 PLN
662-7379 662-7375 662-7367 662-7059 662-7386 662-7377 662-7378 662-7383 662-7373 662-7369 WTWA106649
Ultra-Micro Cells do pomiarów fluorescencji
Elektrody Elektrody jonoselektywne
These easy-to-use, robust, high quality ion-selective electrodes are ideal for field applications.
  • Combined with built-in reference electrodes which make them particularly suitable for measurement of small volume samples
Informacje do zamówienia: These ion selective electrodes will require additional electrolyte and ion strength adjustment solutions (ISA) as well as freshly prepared standards (see ISA solutions).