Drukuj
Kasetki biopsyjne do przechowywania i zanurzania, Histosette ® I M499
Dostawca: Simport Scientific
M499-6 M499-12 M499-10 M499-11 M499-2 M499-3 M499-4 M499-5 M499-7 M499-8 M499-9
720-0946EA 1090 PLN
720-0946 720-1538 720-0940 720-0941 720-0942 720-0943 720-0944 720-0945 720-1542 720-0947 720-0948
Kasetki biopsyjne do przechowywania i zanurzania, Histosette ® I M499
Kasety do osadzania Kasety biopsyjne
Acetal polymer. Disposable cassettes similar to M498 series but specially designed to hold biopsy specimens during the embedding process.
  • No need for biopsy pads
  • Square openings (1 mm) to maximise fluid exchange and to ensure proper drainage
  • Integral lid
  • 45° angle writing area to enable use with some automated cassette-labelling instruments