Drukuj
Pincety powlekane PTFE,
Dostawca: BOCHEM
1800 1801 1802
232-0013EA 197 PLN
232-0013 BOCH1801 232-0015
Pincety powlekane PTFE,
Pincety
Stainless steel, PTFE coating.
  • Anti-magnetic
  • Very high chemical and thermal resistance
  • Smooth surface prevents any contamination and makes cleaning easier