Drukuj
Ciśnieniowa jednostka filtracyjna, Sartobran®-P
Dostawca: Sartorius
5235306D8-OO-A 5235307H7-OO-A 5235307H9-SS-A 5235307H8-SO-A 5235306D9-OO-A 5235306D9-SS-A 5235307H0-SS-V 5235307HS-FF-M 5235307H7-HH-A 5232107H1----P
515-0162EA 3780 PLN
515-0162 515-0164 515-0259 515-0256 515-0163 515-0250 515-0253 515-0260 SARP5235307H7-HH-A SARP5232107H1----P
Ciśnieniowa jednostka filtracyjna, Sartobran®-P
Filtry Kapsuły filtracyjne z membraną
For filtration of protein containing solutions. Compatible with aqueous solutions of pH 4 to 8 and most alcohols and hydrocarbons.
  • Integrity tested
PP housing, double layer cellulose acetate membrane, autoclavable.
Max. pressure: 4 bar at 20 °C.
Also available with sanitary flange on request.
Also available with sanitary flange on request.