Drukuj
System wielostanowiskowy do filtracji próżniowej, Combisart®
Dostawca: Sartorius
Combisart®
516-7030EA 786 PLN
516-7030 515-0188 611-1247
System wielostanowiskowy do filtracji próżniowej, Combisart®
Komory próżniowe
The Combisart® system enables you to select the optimal hardware and consumables for your needs in quality assurance. It features a modular design and field proven standard accessories to make your choice easier.
At the heart of the system is a stainless steel manifold designed to accomodate all types of filter holders and funnels such as:
- ready to use units, BioSart® 100 monitors and BioSart® 250 funnels
- stainless steel funnels that can be flame sterilised and used for colony counting
Autoclavable polycarbonate and glass filter holders
Dodatkowe informacje: Manifold with funnels, lids, taps and connectors.