Drukuj
Osłony twarzy, Relrsi/Relpsi
Dostawca: Bollé Safety
RELRSI RELPSI
111-0334EA 248 PLN
111-0334 111-1012
Osłony twarzy, Relrsi/Relpsi
Osłony twarzy
Protective face shield equipped with adjustable headgear to ensure that the visor remains securely in place.
  • Panoramic vision
  • Ultra-resistant, flip-up face shield, mounted in yellow PP
  • Screen made from 1,5 mm thick fog resistant acetate (model Relrsi) or from 1,0 mm thick PC (model Relpsi)
  • Acetate face shield provides protection against short circuit electric arcs
Weight: 200 g
EN 166, EN 170 (Acetate face shield only)
Certyfikaty: EN 166, EN 170 (acetate face shield only).