Drukuj
Zatyczki do uszu na pałąku, 1310
Dostawca: 3M
1310 1311 ES01300 ES01301 ES01300UK
111-1031EA 567 PLN
111-1031 111-1032 111-0445 111-0446 111-1688
Zatyczki do uszu na pałąku, 1310
Zatyczki do uszu Zatyczki do uszu
These earplugs are ideal for long periods of use with changing noise levels.
  • Flexible band design helps maintain a constant yet comfortable low pressure irrespective of head size
  • Can be worn behind the head or under the chin
  • Round plugs do not enter the ear canal for better hygiene
  • Replacement plugs are economical and easy to fit
SNR: 26 dB
EN 352-2
Certyfikaty: EN 352-2
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable