Drukuj
Jednorazowe zatyczki do uszu, 1100/1110
Dostawca: 3M
1100 1110
111-1021EA 326 PLN
111-1021 111-1022
Jednorazowe zatyczki do uszu, 1100/1110
Zatyczki do uszu Zatyczki do uszu, jednorazowe
Soft, smooth, dirt resistant foam earplugs mould to the unique shape of each ear for excellent hearing protection.
  • Bright orange to enhance visibility
  • Can be combined with ear muffs
  • Available with cord for easy removal and retrieval
The tapered shape allows for a comfortable fit for a wide range of ear sizes.
SNR: 37 dB.
EN 352-2
Certyfikaty: EN 352-2.
Uwaga: * One size fits all.