Drukuj
Parownice Płytkie
Dostawca: MORGAN ADVANCED MATERIALS HALD
L888/0 L888/00 L888/000 L888/10 L888/2 L888/4 L888/6 L888/6A L888/8 L888/9
216-1211EA 82.3 PLN
216-1211 216-1210 216-4285 216-1218 216-1212 216-1213 216-1214 216-1215 216-1216 216-1217
Parownice Płytkie
Tace/Naczynia/Parownice Parownice
Porcelain, glazed.
  • Very good chemical resistance
  • Heat resistant up to 1000 °C
  • Low thermal expansion
For a wide range of laboratory applications. With spout and flat bottom. Glazed white except for the outside bottom surface.
EN 60672-3 Group C100, Type C110
Certyfikaty: EN 60672-3 Group C100, Type C110