Drukuj
Eksykatory szafkowe, SICCO seria Mini Basic
Dostawca: Bohlender
V1850-01 V1850-02 V1850-03
467-4105EA 755 PLN
467-4105 467-4106 467-0051
Eksykatory szafkowe, SICCO seria Mini Basic
Eksykatory Eksykatory szafkowe
Clear, polycarbonate. Each compartment has one door with snap fit closing and integrated gasket ensuring easy opening and tight closure. Temperature resistant from −35 to +70 °C, suitable for use under normal pressure.
  • Large opening and door with wide opening angle enable user to access the entire interior space
  • Robust construction, lightweight, portable and stackable
  • Supplied with desiccant which is in a handy, resealable container
Informacje do zamówienia: Shelves must be ordered separately.