Drukuj
Kuwety jednorazowe, UVette®
Dostawca: EPPENDORF
0030106.300 4099100.007 0030106.318
634-1921EA 380 PLN
634-1921 634-1922 634-1923
Kuwety jednorazowe, UVette®
Kuwety Kuwety spektrofotometryczne
Konstrukcja kuwety ma podstawę w kształcie lejka, która zapobiega efektowi kapilarnemu i zapewnia, że ciecze stale pozostają w środku obszaru pomiarowego.
  • Compatible with most spectrophotometers with the appropriate adapter
  • Four optical windows and graduations at 500 and 1000 µl
  • No adapters required for use with Eppendorf BioPhotometer
The design allows for a 2 and 10 mm optical path length by rotation of the UVette® by 90°. Using the 2 mm path length, there is in most cases no need for dilutions of highly concentrated samples.
Routine pack: Bulk cuvettes in a reclosable box.
Starter set: 80 individually packed UVettes with universal adapter for a 15 mm light centre height (including GeneQuant) convertible to 8,5 mm.
Sposób pakowania: Individually blister-packed for contamination-free work, DNA, RNase and protein-free or bulk cuvettes in a reclosable box.