Drukuj
Kolumny HPLC, skala preparatywna, Hibar®
Dostawca: MERCK PRODUCTION CHEMICALS
1.51482.0001 1.51484.0001 1.51485.0001 1.51478.0001
1.51482.0001EA 0 PLN
1.51482.0001 1.51484.0001 1.51485.0001 1.51478.0001
Kolumny HPLC, skala preparatywna, Hibar®
Kolumny chromatograficzne
Manufactured under strictly controlled conditions to ensure both excellent results and reproducibility. Each column is supplied with a Certificate of Analysis.
These columns are ready to use and can be easily connected to any HPLC system. Methods developed using a particular sorbent at analytical scale can be directly scaled up to preparative scale, for problem-free use.