Drukuj
Kolumny kapilarne, BPX70
Dostawca: Trajan Scientific and Medical
054600 054602 054603 054606 054607 054612 054613 054616 054617 054620 054621 054622 054623 054624
568-0170EA 2250 PLN
568-0170 558-0332 558-0333 558-0334 568-0172 568-0174 568-0175 568-0176 568-0177 568-0179 568-0180 558-0252 558-0335 568-0181
Kolumny kapilarne, BPX70
Kolumny chromatograficzne
Polarity has been designed for fatty acid methyl ester (FAME) analysis.
70% cyanopropyl polysilphenylene-siloxane.
Max. temperature: 260 °C
Suitable replacement for: CP-Sil88, DB-23, HP-23, Rtx-2330, SP-2330, SP-2380.