Drukuj
Kombinezon, Mutex 2
Dostawca: HONEYWELL SAFETY
4180002/S 4180002/XXXL
113-1662EA 0 PLN
113-1662 113-1667
Kombinezon, Mutex 2
Odzież jednorazowa
Anti-static properties inside and out allow this garment to be worn even for activities subject to very stringent requirements (cleanroom production, micro-electronics, hygiene measures). The fleece with its special membrane makes the overall extremely water-repellent.
  • Water-repellent breathable membrane offers optimum comfort, even when worn for long periods
  • No seams on arms and shoulders for extra protection
  • With integrated placket over extra-long zip (820 mm)
  • Anti-static surface, inside and outside
  • Hood and elasticated wrists and ankles
Colour: White
Certyfikaty: Type 5/6, EN 13034, EN 14126, EN 1149-1, Cat. III