Drukuj
Ochronniki na obuwie, Secutex® pro, PP/PE
Dostawca: Zellstoff-Vertriebs-GmbH & Co. KG
30250-90 30254-90
ZVGA30250-90EA 561 PLN
ZVGA30250-90 ZVGA30254-90
Ochronniki na obuwie, Secutex® pro, PP/PE
Odzież jednorazowa
These shoe covers protect against aggressive chemicals and other substances.
  • Lint-free, resistant to abrasion, tear and water
  • Permeable to air and water vapour ensures breathability
  • Anti-static
*One size fits all.