Drukuj
VWR®, Fartuchy dla gości, PP, jednorazowe, proste mankiety
Dostawca: VWR Collection
113-8276EA 622 PLN
113-8276 113-8277 113-8275
VWR®, Fartuchy dla gości, PP, jednorazowe, proste mankiety
Odzież jednorazowa
These disposable laboratory coats made from non woven PP feature a tapered collar and one breast pocket. Ideal for non critical applications.
  • Bound seams
  • Straight wrist
  • Press stud fastening
  • One pocket
Length: 1020 mm; 870 mm (sleeves)
Weight: 40 g/m²
Colour: white