Drukuj
PVC fartuchy robocze
Dostawca: Elbe-Werkstätten GmbH
090-003 090-004 090-007 090-025
113-3302EA 275 PLN
113-3302 113-3303 113-3305 113-3309
PVC fartuchy robocze
Odzież robocza
The PVC apron has sealed seams and features a strong halter neck and side ties made from PE.
  • Resistant to most inorganic acids and alkalis
  • Durable and tear-resistant
  • Easy to clean, machine washable up to 60 °C
  • Available in different sizes, colours, materials and material strengths
Thickness: 0,3 or 0,5 mm
Colour: white/black