Drukuj
Fartuchy odporne na zimno, Cryo-Apron®
Dostawca: TEMPSHIELD
CA48 CA36 CA42 CA54 CI-A42 P-CA36
113-2602EA 1330 PLN
113-2602 113-2600 113-2601 113-2603 113-1599 TEMPP-CA36
Fartuchy odporne na zimno, Cryo-Apron®
Odzież robocza
Cryo-Aprons® are multi-layer insulated aprons designed to provide protection to the torso and legs from contact with ultra cold surfaces in cryogenic atmospheres and from splashes.
  • Protects against temperatures from −160 to +150 °C
  • Fitted with adjustable straps and quick release buckles at the neck and waist
  • Waterproof nylon outer shell protects from cryogenic splashes
  • Cryo-Industrial® model features a rugged waterproof Cordura® nylon outer shell for improved safety
Ideal for working in deep-freeze environments, with cryogenic liquids, dry ice, liquid nitrogen (vapour phase) or when working with autoclaves. The apron is insulated with a breathable membrane laminated to the inner surface. A flexible non woven scrim is bonded to each side.
EN 511; Cat. III
Certyfikaty: EN 511; Cat. III